NANOSPHERE SIZE STANDARD 100 NM 1 * 15 mL
POSS3100A
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Disposable cuvettes
: Brand
pH buffers, ready to use
: WTW
Acetic acid 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP
: Honeywell Chemicals