Order Entry
Austria
Orders LinkContactUsLinkComponent
Histo/Zyme

Histo/Zyme

50 mL
Catalog # H3292-50ML
Histo/Zyme
Catalog # H3292-50ML
CAS Number:  
Histo/Zyme

50 mL

CAS Number:  
Price
694,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

Histo/Zyme is a stabilized proteolytic enzyme in buffered solution, used to unmask immunoreactive sites altered by fixation and/or the embedding process. Treatment with a proteolytic enzyme in immunohistochemistry (IHC) is required to digest excess linkages and expose the antigen. The use of Histo/Zyme results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin. Moreover, Histo/Zyme requires only a brief incubation at room temperature. Performing antigen recovery with Histo/Zyme will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.

  • Antigen Retriever
Specifications

Specifications

  • Pk:
    50 mL
  • Cat. No.:
    H3292-50ML
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2…8 °C
  • pH:
    7.2
  • Concentration:
    Ready to Use
  • Enzyme name:
    Histo/Zyme

Specifications