Order Entry
Austria
Orders LinkContactUsLinkComponent
Histo/Zyme

Histo/Zyme

15 mL
Catalog # H3292-15ML
Histo/Zyme
Catalog # H3292-15ML
CAS Number:  
Histo/Zyme

15 mL

CAS Number:  
Price
249,00 €
Log in for availability
1 * 15 mL
Product Details & Documents

Histo/Zyme is a stabilized proteolytic enzyme in buffered solution, used to unmask immunoreactive sites altered by fixation and/or the embedding process. Treatment with a proteolytic enzyme in immunohistochemistry (IHC) is required to digest excess linkages and expose the antigen. The use of Histo/Zyme results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin. Moreover, Histo/Zyme requires only a brief incubation at room temperature. Performing antigen recovery with Histo/Zyme will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.

  • Antigen Retriever
Specifications

Specifications

  • Pk:
    15 mL
  • Cat. No.:
    H3292-15ML
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2…8 °C
  • pH:
    7.2
  • Concentration:
    Ready to Use
  • Enzyme name:
    Histo/Zyme

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.