Toggle drums
300 L, 710 mm, 800 mm
SAMPA926-300
These stainless steel containers from 2 litres up to 300 litres are designed for storing and processing powders and liquids.
- Splash-tight lid secured by toggle clamps
- Hygienic solid rim
- With two side handles
- Fully welded, crevice free interior
- Finished to a high quality mirror polish
- Removable, easy clean silicone seal
Seal is food grade and meets all the latest FDA compliance standards.
- Ø opening:710 mm
- Capacity:300 L
- Height:800 mm
- Seq:07
Accessories
- Description
- Description
- Description
- Description
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



