ContactUsLinkComponent
K2edta sep tubes are for the examination of edta plasma for molecular diagnostics and for viral load determinations. 1 * 1.200 items
Catalog # GREI454235_1200
Supplier:  Greiner Bio-One
K2edta sep tubes are for the examination of edta plasma for molecular diagnostics and for viral load determinations. 1 * 1.200 items
Catalog # GREI454235_1200
Supplier:  Greiner Bio-One
Frequently Bought Together