ContactUsLinkComponent
For recapping opened vacuette® tubes. snap caps are suitable for all 13mm vacuette® tubes. 1 * 5.000 items
Catalog # GREI371527_5000
Supplier:  Greiner Bio-One
For recapping opened vacuette® tubes. snap caps are suitable for all 13mm vacuette® tubes. 1 * 5.000 items
Catalog # GREI371527_5000
Supplier:  Greiner Bio-One
Frequently Bought Together