For recapping opened vacuette® tubes. snap caps are suitable for all 13mm vacuette® tubes. 1 * 5.000 items
Catalog # GREI371522_5000
Supplier: Greiner Bio-One
For recapping opened vacuette® tubes. snap caps are suitable for all 13mm vacuette® tubes. 1 * 5.000 items
Catalog # GREI371522_5000
Supplier: Greiner Bio-One
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals