Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic Phase objective UC PL Ph 20×/0,45 (WD=0,8 mm), Ph2, 0,8 mm
Catalog # MOTI1101001706491
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001706491
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic Phase objective UC PL Ph 20×/0,45 (WD=0,8 mm), Ph2, 0,8 mm
Price
252,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    Ph2 20x/0,45
  • Description:
    CCIS® Plan achromatic Phase objective UC PL Ph 20×/0,45 (WD=0,8 mm), Ph2
  • Seq:
    10
  • Working distance:
    0,8 mm