Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL NCG 100×/1,25/S-Oil (WD=0,16 mm), 0,16 mm
Catalog # MOTI1101001706551
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001706551
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL NCG 100×/1,25/S-Oil (WD=0,16 mm), 0,16 mm
Price
203,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    100x/1,25 (i)
  • Description:
    CCIS® Plan achromatic objective UC PL NCG 100×/1,25/S-Oil (WD=0,16 mm)
  • Seq:
    16
  • Working distance:
    0,16 mm