ContactUsLinkComponent
Beta Tool-2. Stainless Steel Tubing straightener with soft grips for tight spaces. Designed for service engineers, used to remove ki nks or bends in standard 1/16” and 1/8” OD HPLC stainless steel tubing. . 1 * 1 items
Catalog # MICS46020-BT
Supplier:  MICROSOLV
undefined
Beta Tool-2. Stainless Steel Tubing straightener with soft grips for tight spaces. Designed for service engineers, used to remove ki nks or bends in standard 1/16” and 1/8” OD HPLC stainless steel tubing. . 1 * 1 items
Catalog # MICS46020-BT
Supplier:  MICROSOLV
Frequently Bought Together