Order Entry
Austria
Orders LinkContactUsLinkComponent
Containment berms, QuickBerm®

Containment berms, QuickBerm®

Rigid-lock, folded dimensions (610×610×127 mm), 12,7 kg, 662 l, 1800×1200×305 mm
Catalog # 119-0364
Containment berms, QuickBerm®
Catalog # 119-0364
Containment berms, QuickBerm®
Containment berms, QuickBerm®

Rigid-lock, folded dimensions (610×610×127 mm), 12,7 kg, 662 l, 1800×1200×305 mm

Price
735,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Ergonomic design provides 100% inside berm accessibility with an unobstructed side-to-side floor space for vehicle tires, foot traffic, and cleaning.

  • Single-piece construction patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
  • Quick and easy setup, no assembly required
  • Durable EnGuard™V, modified PVC coated fabric withstands long-term UV exposure
  • It can withstands a broad range of chemicals including petroleum, oils, grease, and most acids—durable enough for extended wear
  • Helps comply with EPA for containment and spill prevention (SPCC)

Exceptionally strong structural integrity includes a reinforced CriticalCorner™ design featuring wraparound diagonal radio frequency welds to prevent billowing, unwrapping, or sagging of the corners during full containment for fluid-tight reliability. Sidewalls lower quickly for easy drive-in and out. Portable design folds down quickly for storage and transport.

The built-in Rigid-Lock wall supports maximize usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter. The device only folds down one way, inward towards the inside of the berm. The one-piece design folds down flat and remains integral to the sidewall.

Specifications
  • Capacity:
    662 l
  • Description:
    Rigid-lock, folded dimensions (610×610×127 mm), 12,7 kg
  • Seq:
    01
  • W×D×H:
    1800×1200×305 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.