Fluo-4 has its absorption maximum at 494 nm, thus making it excitable by the argon-ion laser. Fluo-4 is essentially nonfluorescent w ithout Ca2+ present, but the fluorescence increases at least 100 times on Ca2+ binding. 1 * 0,5 mg
Catalog # 50019.
Supplier: Biotium
Frequently Bought Together