BISPHENOL S 1 * 100 mg
Catalog # EHERC10655940
Supplier: EHRENSTORFER
Frequently Bought Together
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
VWR®, Flash point reference standard, Pensky Martens
Supplier: VWR Chemicals
Elemental analysis standards, sulphur standards, VHG
Supplier: LGC Standards PROMOCHEM
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
2,3,4,6-Tetrachlorophenol analytical standard, Supelco®
Supplier: Merck
Mecoprop certified 1 * 0,1 g
Supplier: EHRENSTORFER
Iothalamic Acid, Reference Standard
Supplier: USP
Indene
Supplier: EHRENSTORFER
Indium Standard for ICP 1 * 100 mL
Supplier: Merck
Supelco® Organic Reference Standard, Nonadecanoic Acid
Supplier: Merck
Terbufos-oxon-sulfoxide 1 * 10 mg
Supplier: EHRENSTORFER
Perindopril Related Compounds, Reference Standard
Supplier: USP
LOSARTAN-D4 1MG 1 * 1 mg
Supplier: LGC Standards PROMOCHEM
Ceriodaphnia dubia, proficiency testing material, Supelco®
Supplier: Merck