Agilent J and W HP-INNOWax GC column, 60 m, 0.25 mm, 0.25 µm, 5 inch cage 1 * 1 items
Catalog # HEWL19091N-136IE
Supplier: Agilent
Agilent J and W HP-INNOWax GC column, 60 m, 0.25 mm, 0.25 µm, 5 inch cage 1 * 1 items
Catalog # HEWL19091N-136IE
Supplier: Agilent
Frequently Bought Together
HPLC columns, SeQuant® ZIC®-pHILIC
Supplier: Merck
Capillary bottle, CERTAN®
Supplier: Merck
XELA UHPLC Columns, C18-AQ-PolE
Supplier: ISERA
Accessories for waste tubes, b.safe
Supplier: Bohlender
Accessories for gas filters Super Clean®
Supplier: Restek
ISAspher HPLC Columns, C30-CXT
Supplier: ISERA
Microvolume syringes 25 to 500 µl
Supplier: Trajan Scientific and Medical
GC capillary column connectors, Swagelok®
Supplier: PerkinElmer
Polystyrene standards, GPC/SEC columns calibration
Supplier: VARIAN
Capillary columns, CP-Sil
Supplier: VARIAN
Capillary columns, Rtx®-5 Amine (fused silica)
Supplier: Restek
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
Diamond RSH/RTC syringes
Supplier: Trajan Scientific and Medical
Chromatography Tubing, PTFE
Supplier: Avantor
Crimp Caps for Crimp Neck Vials, N 11
Supplier: MACHEREY-NAGEL
ISAspher HPLC Columns, C18 BDS
Supplier: ISERA