ContactUsLinkComponent
Agilent J and W GC packed column in UltiMetal tubing, length 6.56 ft (2 m), 1/8 inch od, 2 mm id, ShinCarbon ST packing, mesh size 5 0/80, preconditioned 1 * 1 items
Catalog # HEWLG3591-81187
Supplier:  Agilent
undefined
Agilent J and W GC packed column in UltiMetal tubing, length 6.56 ft (2 m), 1/8 inch od, 2 mm id, ShinCarbon ST packing, mesh size 5 0/80, preconditioned 1 * 1 items
Catalog # HEWLG3591-81187
Supplier:  Agilent
Frequently Bought Together