RF312DUAL WiFi Datalogger High Accuracy + 12-months FREE Cloud Application, large LCD, External Temperature Only, 2x 2m External Probe included. Including wall bracket and micro USB Lead. -40°C to +125°C (-40°F to +257°F) 1 * 1 items
Catalog # COMARF312DUALPLUS
Supplier: COMARK
Frequently Bought Together