Orders LinkContactUsLinkComponent
Graphite tubes for atomic absorption
Pyrocoated Zeeman shroud GTA120
Catalog # PERCN9304634
Supplier:   PerkinElmer
Graphite tubes for atomic absorption
Catalog # PERCN9304634
Supplier:  PerkinElmer
Graphite tubes for atomic absorption
Pyrocoated Zeeman shroud GTA120
Price
144,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Graphite tubes made with highest quality material.
Suitable AA instruments.
Specifications
  • Description:
    Pyrocoated Zeeman shroud GTA120
  • Seq:
    512
  • Cat. No.:
    PERCN9304634