Graphite tubes for atomic absorption
Pyrolytically coated partition graphite tube for extended life
Catalog # PERCN9304627
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Graphite tubes made with highest quality material.
Suitable AA instruments.
Specifications
- Description:Pyrolytically coated partition graphite tube for extended life
- Seq:505