Graphite tubes for atomic absorption
Pyrolytically coated (90° contact cone) standard graphite tube
Catalog # PERCN9304612
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Graphite tubes made with highest quality material.
Suitable AA instruments.
Specifications
- Description:Pyrolytically coated (90° contact cone) standard graphite tube
- Seq:301