Graphite tubes for atomic absorption
Pyrolytically coated partition graphite tube
Catalog # PERCN9304625
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Graphite tubes made with highest quality material.
Suitable AA instruments.
Specifications
- Description:Pyrolytically coated partition graphite tube
- Seq:903