Orders LinkContactUsLinkComponent
Graphite tubes for atomic absorption
Graphite tubes for atomic absorption
Bone platform pyrolytically coated tubes for use in plateau tubes
Catalog # PERCN9304624
Supplier:   PerkinElmer
Graphite tubes for atomic absorption
Catalog # PERCN9304624
Supplier:  PerkinElmer
Graphite tubes for atomic absorption
Bone platform pyrolytically coated tubes for use in plateau tubes
Price
313,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Graphite tubes made with highest quality material.
Suitable AA instruments.
Specifications
  • Description:
    Bone platform pyrolytically coated tubes for use in plateau tubes
  • Seq:
    502