ContactUsLinkComponent
CF647 methyltetrazine reacts with cyclooctene (TCO, methylcyclopropene) labeled molecule via copper free click chemistry. This coppe r free bioorthogonal reaction allows staining the surface of live cells or having concerns about native protein funct 1 * 1 mg
Catalog # 96035.
Supplier:  Biotium
CF647 methyltetrazine reacts with cyclooctene (TCO, methylcyclopropene) labeled molecule via copper free click chemistry. This coppe r free bioorthogonal reaction allows staining the surface of live cells or having concerns about native protein funct 1 * 1 mg
Catalog # 96035.
Supplier:  Biotium
Frequently Bought Together