Order Entry
Austria
Orders LinkContactUsLinkComponent
Scroll vacuum pump

Scroll vacuum pump

Scroll Pump
  LBCP7587000
 :   LABCONCO
Scroll vacuum pump
  LBCP7587000
 :  LABCONCO
Scroll vacuum pump
Scroll vacuum pump

Scroll Pump

 
17 700,00 €
   
1 * 1 ST
 

Scroll Vacuum Pumps are low maintenence, corrosion resistent pumps that require no oil.

  • Scrolls be changed every 40000 hours of use
  • Eliminating the oil eliminates hydrocarbon contamination

Handle water vapor better than most types of pumps and make significantly less noise during operation. Works with FreeZone triad, 1, 2, 4, 5, 6, 8, 12 and 18 L freeze dryers.

 
  • Description:
    Scroll Pump
  • Seq:
    01
  • Cat. No.:
    LBCP7587000
  • Ultimate vacuum:
    7 x 10-3 mBar (Partial Pressure)
  • Dimensions:
    304,8L×508W×381H mm
  • Final vacuum:
    <7×10-3 mbar
  • Electrical:
    115V, 50/60Hz
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.