Orders LinkContactUsLinkComponent
Brushes, FlexBrush™
Brushes, FlexBrush™
S-Brush adapter, S-shaped brush, stainless steel with plastic handle, Nylon, 57,1 mm, Stainless steel, 533 mm
Catalog # MICRBRWM-22512S_U
Supplier:   MICRONOVA
Brushes, FlexBrush™
Catalog # MICRBRWM-22512S_U
Supplier:  MICRONOVA
Brushes, FlexBrush™
Brushes, FlexBrush™
S-Brush adapter, S-shaped brush, stainless steel with plastic handle, Nylon, 57,1 mm, Stainless steel, 533 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
  • Bendable brush with polyester or microfibre cover
  • Acme threaded end attaches to a variety of handles for extended reach
  • Threaded handle (STA series) can be added for extended reach
  • Covers are compatible with strong disinfectants and autoclavable
Specifications
  • Bristle material:
    Nylon
  • Brush head Ø:
    57,1 mm
  • Handle material:
    Stainless steel
  • Description:
    S-Brush adapter, S-shaped brush, stainless steel with plastic handle
  • Total length:
    533 mm