ContactUsLinkComponent
Biotinylated Human OX40, Avitag,His Tag (recommended for biopanning) (TN4-H82E4) is expressed from human 293 cells (HEK293). It cont ains AA Leu 29 - Ala 216 (Accession NP003318.1). 1 * 200 µG
Catalog # ACRBTN4-H82E4200UG
Supplier:  ACRO Biosystems
Biotinylated Human OX40, Avitag,His Tag (recommended for biopanning) (TN4-H82E4) is expressed from human 293 cells (HEK293). It cont ains AA Leu 29 - Ala 216 (Accession NP003318.1). 1 * 200 µG
Catalog # ACRBTN4-H82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together