SEAL & O-RING FOR P200 SET OF 5 1 * 5 items
Catalog # GILSF144865
Supplier: GILSON
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Pipette tips
Supplier: Sartorius
SEAL & O-RING FOR P1000 SET OF 5 1 * 5 items
Supplier: GILSON
pH electrodes, SenTix® PLUS
Supplier: WTW
Sodium azide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Hexamethylenetetramine ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Cleaning swab
Supplier: KOEHLER TECHNISCHE PRODUKTEN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
SEAL & O-RING FOR P100 SET OF 5 1 * 5 items
Supplier: GILSON
SERVICE KIT 1ST LEVEL, P10ML 1 * 1 KIT
Supplier: GILSON