CSPW2 WASH BUFFER, CONCENTRATE 1 * 200 mL
Catalog # CSPW2-200
Supplier: OMEGA BIO-TEK
Frequently Bought Together
Serological pipettes, Stripette™
Supplier: Corning
RNase A
Supplier: OMEGA BIO-TEK
Proteinase K solution
Supplier: PanReac AppliChem
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9% for molecular biology
Supplier: PanReac AppliChem
cDNA synthesis kits, qScript™
Supplier: Quantabio
Mag-Bind® Plant DNA DS 96 kit
Supplier: OMEGA BIO-TEK
Phusion® Green High-Fidelity DNA Polymerase
Supplier: FINNZYMES REAGENTS
Nucleobond Bottle Top Filter Type 2 (5) 1 * 1 KIT
Supplier: MACHEREY-NAGEL
Buffer CF (1000 mL) Lysis Buffer CF bottle of 1000 mL 1 * 1.000 mL
Supplier: MACHEREY-NAGEL
illustra™ TempliPhi™100/500 Amplification Kits
Supplier: Cytiva
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
assay complete thawing reagent T3 (2 x 20 ml) 1 * 100 mL
Supplier: DISCOVERX
Mag-Bind® RXNPure Plus kit
Supplier: OMEGA BIO-TEK
FastAP Thermosensitive Alkaline Phosphatase 5x1000 units (0,1u/µl) 1 * 1 KIT
Supplier: Thermo Fisher Scientific
Multiple well cell culture plates, Costar®
Supplier: Corning