Boxes, with Hinged Lid
50×50×18 mm
Catalog # PLANG3957
Marketsource
Supplier: PLANO W. PLANNET
Jump to:
Product Details & Documents
These clear boxes are made of polystyrene.
- With hinged lid
- Good chemical resistance
- Light and stackable
Multipurpose boxes for a wide range of storage and transport applications.
Specifications
- Seq:02
- W×D×H:50×50×18 mm