COLOUR STANDARD USP FERRIC CHLORIDE CS 1 * 100 mL
Catalog # REGEUSPCS102
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Volumetric flasks, class A, white markings, BLAUBRAND®
Supplier: Brand
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Certified headspace crimp top vials
Supplier: Agilent
Phenolphthalein 1% in ethanol 70% pH-indicator
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Citric acid, anhydrous 99.5-100.5%, powder Ph. Eur.
Supplier: VWR Chemicals
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals