COLOUR STANDARD USP CUPRIC SULFATE CS 1 * 100 mL
Catalog # REGEUSPCS101
Supplier: REAGECON
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
COLOUR STANDARD USP FERRIC CHLORIDE CS 1 * 100 mL
Supplier: REAGECON
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® Economy Line 2.0, Laboratory Chairs
Supplier: VWR Collection