Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids, Haake

Bath fluids, Haake

Synth 260, +50...+ 250 °C, 275 °C, 260 °C, 140 cP, Yellow, 10 L
Catalog # HAKA9990214
Supplier:   THERMO ELECTRON LED
Bath fluids, Haake
Catalog # HAKA9990214
CAS Number:  
Bath fluids, Haake

Synth 260, +50...+ 250 °C, 275 °C, 260 °C, 140 cP, Yellow, 10 L

CAS Number:  
Price
636,00 €
Log in for availability
1 * 10 L
Product Details & Documents

These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.

  • Silicone oils carry a low risk of inflammation, do not give off unpleasant odours and have a long service life
  • Low corrosive tendency

The oils are not suitable for contact with silicone tubing.

Documents

Specifications

Specifications

  • Pk:
    10 L
  • Colour:
    Yellow
  • Fire point:
    275 °C
  • Flash point:
    260 °C
  • Temperature range:
    +50...+ 250 °C
  • Type:
    Synth 260
  • Viscosity:
    140 cP

Specifications