Pipe Systems, FlexTube and FlexTubePro, SymLine®
Product Details & Documents
These flexible tubes made from PTFE can be used in safety cabinets observing the specified maximum dimension for drillings. They are the ideal solution for installation in confined spaces since they can be placed easily into clearances and provide the flexibility required in safety cabinets with pull-out containers.
- Made of PTFE for best possible chemical resistance
- Adjustable and easy to install; shorten tube as required, then mount the optimized screw connection and sealing
- Electrostatic conductive <106 (max) Ω
- 16,9 or 19,8 mm Ext.Ø
- Available in 4 different standard lengths: 600, 1200, 1500, and 2500 mm
FlexTube with inner wall spiral or or FlexTubePro Inner wall smooth. The two most flexible and adaptable tubing systems for the safe disposal of liquid HPLC waste. Flexible, expandable, chemically resistant PTFE tubing with a large bending radius. SymLine® FlexTube Pro with unruffled inner wall prevents waste and residues in tubes.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Tubing ext.Ø:16,9 mm
- Description:SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
- Length:2500 mm
- Seq:08
Accessories
Product Information
- Catalog #AvailabilityPrice
- Tube holder for Symline FlexTube Pro, suitable for all cabinetsTube holder for Symline FlexTube Pro, suitable for all cabinetsSCTV106558Log in for availability1 * 1 ST - 16,38 €
Product Information
- Catalog #AvailabilityPrice
- Tube holder for Symline FlexTube, suitable for all cabinetsTube holder for Symline FlexTube, suitable for all cabinetsSCTV106562Log in for availability1 * 1 ST - 16,38 €
Product Information
- Catalog #AvailabilityPrice
- Screw GL25 for FlexTube, set of 2 pcs.
Product Information
- Catalog #AvailabilityPrice
- Screw GL25 for FlexTube Pro, set of 2 pcs.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



