ContactUsLinkComponent
Universal Transmitter for connection of 2 digital IQ sensors, with PROFIBUS connection, 3 x Relays (Please order separately D-SUB connection ADA/D-SUB 902 888) 1 * 1 items
Catalog # WTWO472111
Supplier:  WTW ONLINE
Universal Transmitter for connection of 2 digital IQ sensors, with PROFIBUS connection, 3 x Relays (Please order separately D-SUB connection ADA/D-SUB 902 888) 1 * 1 items
Catalog # WTWO472111
Supplier:  WTW ONLINE
Frequently Bought Together