Orders LinkContactUsLinkComponent
Face shield
Face shield
Face shield, with helmet adapter, Uncoated, Polycarbonate
Catalog # UVEX9726.014
Supplier:   UVEX
Face shield
Catalog # UVEX9726.014
Supplier:  UVEX
Face shield
Face shield
Face shield, with helmet adapter, Uncoated, Polycarbonate
Price
38,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These polycarbonate and cellulose acetate visors can be attached to helmets by using a length adapter and can be locked in three positions.
  • Robust visor with specially developed adapter for attachment to side helmet slots
  • Extremely lightweight plastic design
  • Material thickness: 1 mm
Ordering information: Visors are supplied with adapter but without helmet. Visors are compatible with helmets uvex airwing.
Specifications
  • Type:
    Uncoated
  • Description:
    Face shield, with helmet adapter
  • Material:
    Polycarbonate
  • Seq:
    02
Complementary Products
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • 111-8232
    Safety helmet, Airwing B
    White
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8235
    Safety helmet, Airwing B
    Yellow
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8216
    Safety helmet, Airwing B
    Orange
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8217
    Safety helmet, Airwing B
    Red
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8218
    Safety helmet, Airwing B
    Green
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8219
    Safety helmet, Airwing B
    Blue
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8234
    Safety helmet, Airwing B-WR
    White
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8237
    Safety helmet, Airwing B-WR
    Yellow
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8253
    Safety helmet, Airwing B-WR
    Orange
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8254
    Safety helmet, Airwing B-WR
    Red
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8255
    Safety helmet, Airwing B-WR
    Green
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8256
    Safety helmet, Airwing B-WR
    Blue
    Log in for availability
    1 * 1 ST - 18,40 €