ContactUsLinkComponent
Cleaning solution elma tec clean S1. Suitable as well for the slightly acid ultrasonic cleaning of non-ferrous and light metals, stainless steels, glass plastics as for the passivation of surfaces of stainless and chromium-containing steels. 1 * 2,5 L
Catalog # ELMA8000162
Supplier:  ELMA SCHMIDBAUER
Cleaning solution elma tec clean S1. Suitable as well for the slightly acid ultrasonic cleaning of non-ferrous and light metals, stainless steels, glass plastics as for the passivation of surfaces of stainless and chromium-containing steels. 1 * 2,5 L
Catalog # ELMA8000162
Supplier:  ELMA SCHMIDBAUER
Frequently Bought Together