Orders LinkContactUsLinkComponent
Multi-packs, Proline® Plus | Avantor
Multi-packs, Proline® Plus
5-pack 10, incl. five single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) and a linear stand
Artikelnr. 613-5736
   Low Manufacturing Impact
Anbieter:   Sartorius
Multi-packs, Proline® Plus
Artikelnr. 613-5736
Anbieter:  Sartorius
   Low Manufacturing Impact
Multi-packs, Proline® Plus
Multi-packs, Proline® Plus
5-pack 10, incl. five single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) and a linear stand
Preis
1 170,00 €
Einloggen für Verfügbarkeit
1 * 1 Pack.
Product Details & Documents
These Multipacks combine a set of Proline® Plus mechanical pipettes with matching tip trays and useful accessories. They offer an opportunity to test and get started with Proline® Plus mechanical pipettes.
  • Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
  • Minimised risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly durable with strengthened structure also for heavier use
It is the widest range of pipettes for various users and applications.
Lieferumfang: All packs include a calibration tool that also acts as a tube opener, as well as the required instructions.
Spezifikationen
  • Description:
    5-pack 10, incl. five single channel pipettes (0,5 - 10 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tip tray boxes (1× 0,1 - 10 µl, 1× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) and a linear stand
  • Seq:
    07
  • Cat. No.:
    613-5736
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact