Push button lockout devices
Lockout bases, small (for 16 mm push buttons)
BRDY130822
: Brady
These lockout devices prevent the unauthorised use of push button emergency devices.
- With standard cover
- Made from clear PC, with translucent red cover
- Suitable for 16, 22 or 30 mm push buttons
- Type:Lockout bases, small (for 16 mm push buttons)
- Seq:09



