Drop-n-Stain CF488A donkey anti-rabbit IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamste 1 * 5 mL
Catalog # BTIU20950
Supplier: Biotium
Frequently Bought Together