Tray Quarz glass in application temperatures of up to 1050°C (maximum 1600°C), 4-sided, translucent, high chemical resistance and purity. Length 324mm, width 152mm, height 35mm. The durability or operating cycles of our quartz glass trays mainly depe 1 * 1 items
Catalog # TSLA73038
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together