Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR®, Digital Mini Block Heaters

VWR®, Digital Mini Block Heaters

Mini block heater**, 1, 0,075 kW, Ambient +5...100 °C, ±0,2 °C, ±0,5 °C
Catalog # 460-0334
   Waste Reducing
Supplier:   VWR Collection
VWR®, Digital Mini Block Heaters
Catalog # 460-0334
Supplier:  VWR Collection
   Waste Reducing
VWR®, Digital Mini Block Heaters
VWR®, Digital Mini Block Heaters

Mini block heater**, 1, 0,075 kW, Ambient +5...100 °C, ±0,2 °C, ±0,5 °C

Price
611,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These mini block heaters are ideal for applications that require accurate, repeatable results. These personal-sized units take up minimal bench space. Choice of model with or without heated lid. Units are ideal for immunoassays, ELISA, LAMP assays, enzyme reactions, denaturations and blood banking.

  • LED display for temperature and time
  • Temperature range from ambient 5 to 100 °C
  • Temperature accuracy: ±0,5 °C (models with heated lid: ±0,3 °C)
  • Microprocessor controlled with temperature calibration via external thermometer or temperature device
  • Timer range: 1 second to 160 hours, or continuous mode
  • Heated lid helps to reduce condensation within sample tube
  • Accommodates interchangeable mini heating blocks for tubes from 0,2 up to 50 ml capacity

Accessories information: Blocks for 15 and 50 ml tubes (460-0342 and 462-0343) cannot be used with the mini block heater with heated lid.

Ordering information: Heating blocks are not included, please order these separately.

Caution: ** EU/CH-plug *** UK-plug

Specifications
  • Heating capacity:
    0,075 kW
  • No. of blocks:
    1
  • Temperature Range:
    Ambient +5...100 °C
  • Seq:
    001
  • Accuracy:
    ±0,5 °C
  • Temp. stability:
    ±0,2 °C
  • Type:
    Mini block heater**
  • Cat. No.:
    460-0334
  • W×D×H:
    127×160×123 mm
  • Environmentally Preferable:
    Waste Reducing
  • Weight:
    1,3 kg