ContactUsLinkComponent
Stericlin® sterilisation pouches, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, 50 x 60 cm, DIN EN ISO 11607 1 * 250 items
Catalog # 129-1292
Stericlin® sterilisation pouches, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, 50 x 60 cm, DIN EN ISO 11607 1 * 250 items
Catalog # 129-1292