Slide micro histobond+s blue adhesive made of soda lime glass of 3rd hydrolytic CL.in compliance with din iso 8037/1 dimensions approx. 76x26 mm, thickness approx. 1 mm with silane treated surfaces with 90 ground edges pre-cleaned autoclavable, 10x2 1 * 2.000 items
Catalog # MARI0810531
Supplier: MARIENFELD
Frequently Bought Together