ContactUsLinkComponent
Short-term tube, DT i-propanol 50/a, standard measuring range of 40 to 4000 ppm 1 * 10 items
Catalog # DRAE8103741
Supplier:  Dräger
Short-term tube, DT i-propanol 50/a, standard measuring range of 40 to 4000 ppm 1 * 10 items
Catalog # DRAE8103741
Supplier:  Dräger
Frequently Bought Together