Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Sample Bags without Closure

200×150 mm, 0,10 mm
Catalog # SCAC2063
Sample Bags without Closure
Catalog # SCAC2063
Sample Bags without Closure
Sample Bags without Closure

200×150 mm, 0,10 mm

Price
331,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Bags made from PE transparent, for the storage or transport of samples.

Available in several sizes.

 : Supplied without closure ties.

Specifications
  • L×W:
    200×150 mm
  • Seq:
    03
  • Thickness:
    0,10 mm