Cream Butyrometer Koehler Gerber Standard Volumetric method for 5 ml cream, with a frosted label on the bulb, ring neck, without a stopper % Fat: 0 - 70 division: 1.0 (± 0.5) 1 * 1 items
Catalog # GEAG03.0318
Supplier: Gerber Instruments AG
Frequently Bought Together