Drum racks
Type 360 with GRP sump pallet (150 l), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 690/1170 mm, 60 kg, 1320×800×1300 mm
Catalog # CEMG7642
Supplier: CEMO
Jump to:
Product Details & Documents
Drum racks for approved and space saving storage of barrels and other containers.
- All drum racks are completely hotdip galvanised and have a corrosion free GRP sump pallet for long-term safe use
- With general construction inspection approval no. Z-40.12-227 granted by DIBt-Berlin
- Universally applicable for waste oil, lyes and acids
Specifications
- Weight:60 kg
- Description:Type 360 with GRP sump pallet (150 l), drum support for 6×60 l barrels and small containers, heights of storage levels from ground 690/1170 mm
- Seq:04
- W×D×H:1320×800×1300 mm
Accessories
Product Information
- Catalog #WeightAvailabilityPrice
- Can holder, galvanised, for drum racks type 120/360/540, with surface 230×110 mmCan holder, galvanised, for drum racks type 120/360/540, with surface 230×110 mmCEMG59172 kgLog in for availability1 * 1 ST - 41,00 €
Product Information
- Catalog #WeightAvailabilityPrice
- Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from groundRotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from groundCEMG76696 kgLog in for availability1 * 1 ST - 235,00 €
Product Information
- Catalog #WeightAvailabilityPrice
- Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drumsDrum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drumsCEMG82541 kgLog in for availability1 * 3 ST - 51,30 €
Product Information
- Catalog #WeightAvailabilityPrice
- Steel grating for drum racks, as 3rd (for type 360) or 4th storage level (for type 540), 1300×620×30 mm, 210 mm storage height from groundSteel grating for drum racks, as 3rd (for type 360) or 4th storage level (for type 540), 1300×620×30 mm, 210 mm storage height from groundCEMG681718 kgLog in for availability1 * 1 ST - 127,00 €
Product Information
- Catalog #WeightAvailabilityPrice
- Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mmCan holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mmCEMG77272 kgLog in for availability1 * 1 ST - 46,80 €
Product Information
- Catalog #WeightAvailabilityPrice
- Drum supports, 3 pieces per storage level, for drum racks type 360/540Drum supports, 3 pieces per storage level, for drum racks type 360/540CEMG78001 kgLog in for availability1 * 1 ST - 48,30 €
Product Information
- Catalog #WeightW×D×HAvailabilityPrice
- Sump pallet, PE, 40 l, without gratingSump pallet, PE, 40 l, without gratingCEMG78715 kg1300×620×80 mmLog in for availability1 * 1 ST - 163,00 €