ContactUsLinkComponent
RING LED MAG. LUMIN. RLLQ 63, 6W/100-240V/50/60HZ/IP20, RF: BLACK, OF: SILVER METALLIC(M4), LENS: 6,0 DPT, BASE, MAINS:2,3 M, PLUG: CEE7/XVI. vendor material number: 113142000-00618824. 1 * 1 items
Catalog # WLDM113142-618824
Supplier:  WALDMANN
RING LED MAG. LUMIN. RLLQ 63, 6W/100-240V/50/60HZ/IP20, RF: BLACK, OF: SILVER METALLIC(M4), LENS: 6,0 DPT, BASE, MAINS:2,3 M, PLUG: CEE7/XVI. vendor material number: 113142000-00618824. 1 * 1 items
Catalog # WLDM113142-618824
Supplier:  WALDMANN
Frequently Bought Together