QUICK COUPLING 1 * 1 items
Catalog # VACU20635851
Supplier: VACUUBRAND
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Accessories for accu-jet® pro
Supplier: Brand
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
MicroWell™ Plates, 96 well, Nunc™
Supplier: Thermo Fisher Scientific
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals