Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue embedding cassettes, mega Type

Tissue embedding cassettes, mega Type

Green
Catalog # KART2925-12
Supplier:   KARTELL
Tissue embedding cassettes, mega Type
Catalog # KART2925-12
Supplier:  KARTELL
Tissue embedding cassettes, mega Type
Tissue embedding cassettes, mega Type

Green

Price
396,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Material: POM. Tissue embedding cassettes Mega Type for samples with big dimensions. Dimension: 28×40×13,6 mm.

Specifications
  • Colour:
    Green