Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue embedding cassettes, mega Type

Tissue embedding cassettes, mega Type

Yellow
Catalog # KART2925-06
Supplier:   KARTELL
Tissue embedding cassettes, mega Type
Catalog # KART2925-06
Supplier:  KARTELL
Tissue embedding cassettes, mega Type
Tissue embedding cassettes, mega Type

Yellow

Price
396,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Material: POM. Tissue embedding cassettes Mega Type for samples with big dimensions. Dimension: 28×40×13,6 mm.

Specifications
  • Colour:
    Yellow
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.